accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1344-43-0

IUPAC Name: oxomanganese
Synonyms: manganese oxide manganese(II) oxide
Molecular Weight (g/mol): 70.94
Molecular Formula: MnO
InChi Key: VASIZKWUTCETSD-UHFFFAOYSA-N
SMILES: O=[Mn]
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (4)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (7)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (4)
  • (2)