accessibility menu, dialog, popup

UserName

Viewing profile as user:
HgS

CAS RN 1344-48-5

IUPAC Name: mercury(2+) sulfanediide
Synonyms: mercury(2+) sulfanediide
Molecular Weight (g/mol): 232.65
Molecular Formula: HgS
InChi Key: LGZXYFMMLRYXLK-UHFFFAOYSA-N
SMILES: [S--].[Hg++]
Physical Form 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)