accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1344-54-3

IUPAC Name: oxo[(oxotitanio)oxy]titanium
Synonyms: titanium(III) oxide
Molecular Weight (g/mol): 143.73
Molecular Formula: O3Ti2
InChi Key: GQUJEMVIKWQAEH-UHFFFAOYSA-N
SMILES: O=[Ti]O[Ti]=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)