accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1344-58-7

IUPAC Name: trioxouranium
Synonyms: uranium trioxide
Molecular Weight (g/mol): 286.03
Molecular Formula: O3U
InChi Key: JCMLRUNDSXARRW-UHFFFAOYSA-N
SMILES: O=[U](=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results