accessibility menu, dialog, popup

UserName

Viewing profile as user:
CaOSiOO

CAS RN 1344-95-2

IUPAC Name: calcium oxosilanebis(olate)
Synonyms: wollastonite
Molecular Weight (g/mol): 116.16
Molecular Formula: CaO3Si
InChi Key: OYACROKNLOSFPA-UHFFFAOYSA-N
SMILES: [Ca++].[O-][Si]([O-])=O
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)