accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13444-24-1

H3CNH(R)HO

CAS RN 13444-24-1

IUPAC Name: (3R)-1-ethyl-3-hydroxypiperidin-1-ium
Synonyms: (3R)-1-ethyl-3-hydroxypiperidin-1-ium
Molecular Weight (g/mol): 130.21
Molecular Formula: C7H16NO
InChi Key: ZNPSUOAGONLMLK-SSDOTTSWSA-O
SMILES: CC[NH+]1CCC[C@@H](O)C1
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results