accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13444-98-9

IUPAC Name: palladium(2+) dihydrate sulfate
Synonyms: palladium(2+) dihydrate sulfate
Molecular Weight (g/mol): 238.51
Molecular Formula: H4O6PdS
InChi Key: TWIRRPLUAGEFNJ-UHFFFAOYSA-L
SMILES: O.O.[Pd++].[O-]S([O-])(=O)=O
Grade 
  • (2)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)