accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13446-19-0

IUPAC Name: magnesium(2+) hexahydrate diperchlorate
Synonyms: magnesium(2+) hexahydrate diperchlorate
Molecular Weight (g/mol): 331.29
Molecular Formula: Cl2H12MgO14
InChi Key: MGBDNCBRZIMSDZ-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.[Mg++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)