accessibility menu, dialog, popup

UserName

Viewing profile as user:
OMoOOOKK

CAS RN 13446-49-6

IUPAC Name: dipotassium dioxomolybdenumbis(olate)
Synonyms: dipotassium molybdate
Molecular Weight (g/mol): 238.14
Molecular Formula: K2MoO4
InChi Key: NUYGZETUIUXJCO-UHFFFAOYSA-N
SMILES: [K+].[K+].[O-][Mo]([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (2)
  • (6)
  • (1)
  • (3)
  • (1)
  • (3)
  • (4)
  • (2)
  • (4)