CAS RN 134470-38-5
IUPAC Name:
1-[(3E)-4-[3-(4-fluorophenoxy)phenyl]but-3-en-2-yl]-1-hydroxyurea
Synonyms:
1-[(3E)-4-[3-(4-fluorophenoxy)phenyl]but-3-en-2-yl]-1-hydroxyurea
Molecular Weight (g/mol):
316.33
Molecular Formula:
C17H17FN2O3
InChi Key:
UAIYNMRLUHHRMF-AATRIKPKNA-N
SMILES:
CC(\C=C\C1=CC=CC(OC2=CC=C(F)C=C2)=C1)N(O)C(N)=O
1
–
1
of
1
results
1
BW-B 70C, R&D Systems™
CAS: 134470-38-5 Molecular Formula: C17H17FN2O3 Molecular Weight (g/mol): 316.332 InChI Key: UAIYNMRLUHHRMF-AATRIKPKSA-N PubChem CID: 5353454 ChEBI: CHEBI:75307 IUPAC Name: 1-[(E)-4-[3-(4-fluorophenoxy)phenyl]but-3-en-2-yl]-1-hydroxyurea SMILES: CC(C=CC1=CC(=CC=C1)OC2=CC=C(C=C2)F)N(C(=O)N)O