accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13450-88-9

IUPAC Name: gallium(3+) tribromide
Synonyms: gallium(III) bromide
Molecular Weight (g/mol): 309.44
Molecular Formula: Br3Ga
InChi Key: SRVXDMYFQIODQI-UHFFFAOYSA-K
SMILES: [Ga+3].[Br-].[Br-].[Br-]
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)