accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13450-91-4

IUPAC Name: gallium(3+) triiodide
Synonyms: gallium(III) iodide
Molecular Weight (g/mol): 450.44
Molecular Formula: GaI3
InChi Key: DWRNSCDYNYYYHT-UHFFFAOYSA-K
SMILES: [Ga+3].[I-].[I-].[I-]
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (6)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)