accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13450-95-8

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 588.31
Molecular Formula: GeH8I4
InChi Key: WTPFKACBVKJUIS-UHFFFAOYSA-N
SMILES: [GeH4].I.I.I.I
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)