accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrBr

CAS RN 1345021-52-4

IUPAC Name: 3-(3,5-dibromophenyl)-9-phenyl-9H-carbazole
Synonyms: 3-(3,5-dibromophenyl)-9-phenylcarbazole
Molecular Weight (g/mol): 477.2
Molecular Formula: C24H15Br2N
InChi Key: GXVXADLFVMCWNC-UHFFFAOYSA-N
SMILES: BrC1=CC(=CC(Br)=C1)C1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results