CAS RN 13451-14-4
CAS RN 13451-14-4
IUPAC Name:
telluric acid dihydrate
Synonyms:
telluric acid dihydrate
Molecular Weight (g/mol):
229.64
Molecular Formula:
H6O6Te
InChi Key:
VCYUGZHEVVXOPD-UHFFFAOYSA-N
SMILES:
O.O.O[Te](O)(=O)=O
1
–
1
of
1
results
1
Telluric acid dihydrate, 99+%
CAS: 13451-14-4 Molecular Formula: H6O6Te Molecular Weight (g/mol): 229.64 MDL Number: MFCD00150387 InChI Key: VCYUGZHEVVXOPD-UHFFFAOYSA-N Synonym: telluric acid dihydrate,telluricacidwhitepowder,telluric acid-te