accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 134524-84-8

IUPAC Name: [2'-(diphenylphosphaniumyl)-[1,1'-binaphthalen]-2-yl]diphenylphosphanium; dichlororuthenium
Synonyms: [2'-(diphenylphosphaniumyl)-[1,1'-binaphthalen]-2-yl]diphenylphosphanium; dichlororuthenium
Molecular Weight (g/mol): 796.67
Molecular Formula: C44H34Cl2P2Ru
InChi Key: YEKBVMDAGDTOQB-UHFFFAOYSA-N
SMILES: Cl[Ru]Cl.C1=CC=C(C=C1)[PH+](C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1[PH+](C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results