accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13454-78-9

IUPAC Name: dicaesium(1+) dioxochromiumbis(olate)
Synonyms: caesium chromate
Molecular Weight (g/mol): 381.8
Molecular Formula: CrCs2O4
InChi Key: BROHICCPQMHYFY-UHFFFAOYSA-N
SMILES: [Cs+].[Cs+].[O-][Cr]([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results