accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13454-88-1

IUPAC Name: copper(2+) dihydrate difluoride
Synonyms: copper(2+) dihydrate difluoride
Molecular Weight (g/mol): 137.57
Molecular Formula: CuF2H4O2
InChi Key: CTRICEWDSISGPV-UHFFFAOYSA-L
SMILES: O.O.[F-].[F-].[Cu++]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results