accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13454-94-9

IUPAC Name: dicerium(3+) trisulfate
Synonyms: cerium(III) sulfate
Molecular Weight (g/mol): 568.4
Molecular Formula: Ce2O12S3
InChi Key: OZECDDHOAMNMQI-UHFFFAOYSA-H
SMILES: [Ce+3].[Ce+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)