accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13455-01-1

IPII

CAS RN 13455-01-1

IUPAC Name: triiodophosphane
Synonyms: phosphorus triiodide
Molecular Weight (g/mol): 411.69
Molecular Formula: I3P
InChi Key: PZHNNJXWQYFUTD-UHFFFAOYSA-N
SMILES: IP(I)I
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)