accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13455-22-6

IUPAC Name: potassium sulfurofluoridate
Synonyms: potassium sulfurofluoridate
Molecular Weight (g/mol): 138.15
Molecular Formula: FKO3S
InChi Key: VXJAYNWISQFORV-UHFFFAOYSA-M
SMILES: [K+].[O-]S(F)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)