accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHC

CAS RN 13461-74-0

IUPAC Name: 9-ethynyl-9H-fluoren-9-ol
Synonyms: 9-ethynylfluoren-9-ol
Molecular Weight (g/mol): 206.24
Molecular Formula: C15H10O
InChi Key: MMZVVJGCZZAWBN-UHFFFAOYSA-N
SMILES: OC1(C#C)C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)