accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13462-88-9

IUPAC Name: nickel(2+) dibromide
Synonyms: nickel(II) bromide
Molecular Weight (g/mol): 218.5
Molecular Formula: Br2Ni
InChi Key: IPLJNQFXJUCRNH-UHFFFAOYSA-L
SMILES: [Ni++].[Br-].[Br-]
Molecular Weight (g/mol) 
  • (5)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (5)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)