accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13462-90-3

IUPAC Name: nickel(2+) diiodide
Synonyms: nickel(II) iodide
Molecular Weight (g/mol): 312.5
Molecular Formula: I2Ni
InChi Key: BFSQJYRFLQUZKX-UHFFFAOYSA-L
SMILES: [Ni++].[I-].[I-]
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)