accessibility menu, dialog, popup

UserName

Viewing profile as user:
ZnSNOSNO

CAS RN 13463-41-7

IUPAC Name: zinc pyrithione
Synonyms: zinc pyrithione
Molecular Weight (g/mol): 317.69
Molecular Formula: C10H8N2O2S2Zn
InChi Key: OTPSWLRZXRHDNX-UHFFFAOYSA-L
SMILES: [Zn++].[O-][N+]1=CC=CC=C1[S-].[O-][N+]1=CC=CC=C1[S-]
Percent Purity 
  • (2)
  • (3)
  • (7)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (7)
  • (2)
Grade 
  • (1)
  • (7)
Physical Form 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (7)
  • (1)
  • (2)
  • (7)
  • (3)
  • (7)
  • (2)
  • (3)
  • (7)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (7)
  • (2)
  • (1)
  • (7)
  • (3)
  • (4)