accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2H2NOHSOHOO

CAS RN 13464-80-7

IUPAC Name: hydrazine; sulfuric acid
Synonyms: hydrazine sulfate
Molecular Weight (g/mol): 130.12
Molecular Formula: H6N2O4S
InChi Key: ZGCHATBSUIJLRL-UHFFFAOYSA-N
SMILES: NN.OS(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results