accessibility menu, dialog, popup

UserName

Viewing profile as user:
ScBrBrBr

CAS RN 13465-59-3

IUPAC Name: scandium(3+) tribromide
Synonyms: scandium bromide
Molecular Weight (g/mol): 284.67
Molecular Formula: Br3Sc
InChi Key: APPHYFNIXVIIJR-UHFFFAOYSA-K
SMILES: [Sc+3].[Br-].[Br-].[Br-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results