accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13465-84-4

IUPAC Name: tetraiodosilane
Synonyms: silicon tetraiodide
Molecular Weight (g/mol): 535.7
Molecular Formula: I4Si
InChi Key: CFTHARXEQHJSEH-UHFFFAOYSA-N
SMILES: I[Si](I)(I)I
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)