accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13465-95-7

IUPAC Name: barium(2+) diperchlorate
Synonyms: barium perchlorate
Molecular Weight (g/mol): 336.22
Molecular Formula: BaCl2O8
InChi Key: OOULUYZFLXDWDQ-UHFFFAOYSA-L
SMILES: [Ba++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
  • (1)
Quantity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (1)
  • (4)
Grade 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)