accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13466-78-9

IUPAC Name: 3,7,7-trimethylbicyclo[4.1.0]hept-3-ene
Synonyms: monoterpenes 3-carene, (R)- delta(sup 3)-carene (r)-3-carene 3-carene, (s)-(cis)-isomer carene 3-carene, (r)- 3-carene (S)-(cis)-isomer 3-carene 3-carene, (r)-isomer
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: BQOFWKZOCNGFEC-UHFFFAOYNA-N
SMILES: CC1=CCC2C(C1)C2(C)C
Percent Purity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)