accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 134678-17-4

IUPAC Name: 4-amino-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-1,2-dihydropyrimidin-2-one
Synonyms: lamivudine epzicom (2s-cis)-isomer lamivudine (2s-cis)-lamivudine 3TC (2S-cis)-lamivudine (-)-2'-deoxy-3'-thiacytidine hepitec lamivudine, (2S-cis)-isomer 3'-thia-2',3'-dideoxycytidine
Molecular Weight (g/mol): 229.25
Molecular Formula: C8H11N3O3S
InChi Key: JTEGQNOMFQHVDC-NKWVEPMBSA-N
SMILES: NC1=NC(=O)N(C=C1)[C@@H]1CS[C@H](CO)O1
Quantity 
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)