accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OH2OThONOOONOOONOOONOO

CAS RN 13470-07-0

IUPAC Name: thorium(4+) tetrahydrate tetranitrate
Synonyms: thorium(4+) tetrahydrate tetranitrate
Molecular Weight (g/mol): 552.11
Molecular Formula: H8N4O16Th
InChi Key: VAVIMIAZQDNXID-UHFFFAOYSA-N
SMILES: O.O.O.O.[Th+4].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)