accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13473-26-2

IUPAC Name: 2',4',5',7'-tetrabromo-4,5,6,7-tetrachloro-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: solvent red 48
Molecular Weight (g/mol): 785.66
Molecular Formula: C20H4Br4Cl4O5
InChi Key: ZYIBVBKZZZDFOY-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C2C(OC3=C(Br)C(O)=C(Br)C=C3C22OC(=O)C3=C(Cl)C(Cl)=C(Cl)C(Cl)=C23)=C1Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results