CAS RN 13473-90-0
IUPAC Name:
aluminium(3+) nonahydrate trinitrate
Synonyms:
aluminium nitrate
Molecular Weight (g/mol):
375.13
Molecular Formula:
AlH18N3O18
InChi Key:
SWCIQHXIXUMHKA-UHFFFAOYSA-N
SMILES:
O.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
1
–
7
of
7
results
1
3
VeriSpec™ Multi-Element Standard 4, Ricca Chemical
5000 ppm: Al, Ca, Mg; 2000 ppm Fe /nManufactured and Tested in an ISO 17025/Guide 34 Facility
4
5
7
VeriSpec™ Primary Interference Analytes, Ricca Chemical
5000 ppm: Al, Ca, Mg; 2000 ppm Fe /nManufactured and Tested in an ISO 17025/Guide 34 Facility