accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONHOH3C

CAS RN 13475-17-7

IUPAC Name: ethyl 2-(4-acetamidophenyl)acetate
Synonyms: ethyl 2-(4-acetamidophenyl)acetate
Molecular Weight (g/mol): 221.26
Molecular Formula: C12H15NO3
InChi Key: OOLUNZSKHFNSCD-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=C(NC(C)=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results