accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOCHHCCHH3CH3CCH3H3CCH3H3CV

CAS RN 13476-99-8

IUPAC Name: vanadium(3+) tris(2,4-dioxopentan-3-ide)
Synonyms: vanadium(3+) triacetylacetonate
Molecular Weight (g/mol): 348.27
Molecular Formula: C15H21O6V
InChi Key: ZVFSRLXFKIWGRT-UHFFFAOYSA-N
SMILES: [V+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results