accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSNFBrBr

CAS RN 1347736-74-6

IUPAC Name: 4,7-dibromo-5-fluoro-2,1,3-benzothiadiazole
Synonyms: 4,7-dibromo-5-fluoro-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 311.95
Molecular Formula: C6HBr2FN2S
InChi Key: KVZDYOVYIHJETJ-UHFFFAOYSA-N
SMILES: FC1=C(Br)C2=NSN=C2C(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results