accessibility menu, dialog, popup

UserName

Viewing profile as user:
LiLiLiOAsOOO

CAS RN 13478-14-3

IUPAC Name: trilithium(1+) arsorate
Synonyms: trilithium(1+) arsenate
Molecular Weight (g/mol): 159.74
Molecular Formula: AsLi3O4
InChi Key: VKJKOXNPYVUXNC-UHFFFAOYSA-K
SMILES: [Li+].[Li+].[Li+].[O-][As]([O-])([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results