accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNNNN

CAS RN 1347814-89-4

IUPAC Name: 1-(cyanomethyl)-2-(pyridin-2-yl)-1H-imidazole-4,5-dicarbonitrile
Synonyms: 1-(cyanomethyl)-2-(pyridin-2-yl)imidazole-4,5-dicarbonitrile
Molecular Weight (g/mol): 234.22
Molecular Formula: C12H6N6
InChi Key: CKIOVYFZWSZUDP-UHFFFAOYSA-N
SMILES: N#CCN1C(=NC(C#N)=C1C#N)C1=CC=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results