accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNSH2NSNH2

CAS RN 1347815-08-0

IUPAC Name: 2-(4-methylphenyl)-1H-imidazole-4,5-dicarbothioamide
Synonyms: 2-(4-methylphenyl)-1H-imidazole-4,5-dicarbothioamide
Molecular Weight (g/mol): 276.38
Molecular Formula: C12H12N4S2
InChi Key: CFNPTMBECAYGCK-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=NC(C(N)=S)=C(N1)C(N)=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results