accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNNNN

CAS RN 1347815-43-3

IUPAC Name: 1-(cyanomethyl)-2-(4-cyanophenyl)-1H-imidazole-4,5-dicarbonitrile
Synonyms: 1-(cyanomethyl)-2-(4-cyanophenyl)imidazole-4,5-dicarbonitrile
Molecular Weight (g/mol): 258.24
Molecular Formula: C14H6N6
InChi Key: OAMIMEALBFOHIE-UHFFFAOYSA-N
SMILES: N#CCN1C(=NC(C#N)=C1C#N)C1=CC=C(C=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results