accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CNH3CH3C

CAS RN 13483-31-3

IUPAC Name: 1,1-dimethyl-2-(propan-2-ylidene)hydrazine
Synonyms: 1,1-dimethyl-2-(propan-2-ylidene)hydrazine
Molecular Weight (g/mol): 100.17
Molecular Formula: C5H12N2
InChi Key: IDSMDKUVIBSETN-UHFFFAOYSA-N
SMILES: CN(C)N=C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results