CAS RN 13485-59-1
IUPAC Name:
1-(2-aminopropanoyl)pyrrolidine-2-carboxylic acid
Synonyms:
proline,D-alanyl
Molecular Weight (g/mol):
186.21
Molecular Formula:
C8H14N2O3
InChi Key:
WPWUFUBLGADILS-UHFFFAOYNA-N
SMILES:
CC(N)C(=O)N1CCCC1C(O)=O