accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3I

CAS RN 134856-58-9

IUPAC Name: [2-(4-iodophenyl)ethynyl]trimethylsilane
Synonyms: [2-(4-iodophenyl)ethynyl]trimethylsilane
Molecular Weight (g/mol): 300.21
Molecular Formula: C11H13ISi
InChi Key: WOOFAKCCTQIPLK-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC1=CC=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results