accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNO

CAS RN 13493-47-5

IUPAC Name: N-(2-formylphenyl)acetamide
Synonyms: N-(2-formylphenyl)acetamide
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: OWMJAQBUFVTERI-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC=C1C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results