accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 135-01-3

IUPAC Name: 1,2-diethylbenzene
Synonyms: diethylbenzenes O-diethylbenzene
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: KVNYFPKFSJIPBJ-UHFFFAOYSA-N
SMILES: CCC1=CC=CC=C1CC
Molecular Weight (g/mol) 
  • (3)
  • (4)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (4)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)