accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOSOHO

CAS RN 135-13-7

IUPAC Name: 2-[(carboxymethyl)sulfanyl]benzoic acid
Synonyms: 2-[(carboxymethyl)sulfanyl]benzoic acid
Molecular Weight (g/mol): 212.22
Molecular Formula: C9H8O4S
InChi Key: GMBZSYUPMWCDGK-UHFFFAOYSA-N
SMILES: OC(=O)CSC1=CC=CC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results