accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNCH3NSN

CAS RN 135-23-9

IUPAC Name: hydrogen N-[2-(dimethylamino)ethyl]-N-[(thiophen-2-yl)methyl]pyridin-2-amine chloride
Synonyms: hydrogen rest-on chloride
Molecular Weight (g/mol): 297.85
Molecular Formula: C14H20ClN3S
InChi Key: BONORRGKLJBGRV-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)CCN(CC1=CC=CS1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results