accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 135-48-8

IUPAC Name: pentacene
Synonyms: pentacene 2,3:6,7-dibenzanthracene lin-dibenzanthracene lin-naphthoanthracene benzo(b)naphthacene
Molecular Weight (g/mol): 278.35
Molecular Formula: C22H14
InChi Key: SLIUAWYAILUBJU-UHFFFAOYSA-N
SMILES: C1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1
Quantity 
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (4)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)