accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 135-49-9

IUPAC Name: 2,7-dimethylacridine-3,6-diamine hydrochloridyl
Synonyms: acridine yellow hydrochloridyl
Molecular Weight (g/mol): 272.76
Molecular Formula: C15H15ClN3
InChi Key: MIVVOLJUKZMTIN-UHFFFAOYSA-N
SMILES: [Cl].CC1=CC2=CC3=CC(C)=C(N)C=C3N=C2C=C1N
Color 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)